3-Bromo-5-methoxypyridine ≥97.0% (by titrimetric analysis)

Supplier: TCI America

B3684-1G B3684-5G
TCB3684-1GEA 56.88 CAD
TCB3684-1G TCB3684-5G
3-Bromo-5-methoxypyridine ≥97.0% (by titrimetric analysis)
3-Bromo-5-methoxypyridine

CAS Number: 50720-12-2
MDL Number: MFCD00234169
Molecular Formula: C6H6BrNO
Molecular Weight: 188.02
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Flash Point (°C): 102
Freezing point (°C): 29

Formula: C₆H₆BrNO
Flash Pt: 102 °C
MDL Number: MFCD00234169
CAS Number: 50720-12-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0d96cdc5a3eb516fd38d8b488321ceb97e8c35a&vid=b0d96cdc5a3eb516fd38d8b488321ceb97e8c35a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR