4-Bromo-o-cresol ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
4-Bromo-2-methylphenol, 1-Bromo-4-hydroxy-3-methylbenzene
B3677-25G
B3677-5G
TCB3677-25GEA
357.08
CAD
TCB3677-25G
TCB3677-5G
4-Bromo-o-cresol ≥98.0% (by GC)
4-Bromo-o-cresol
CAS Number: 2362-12-1
MDL Number: MFCD00055435
Molecular Formula: C7H7BrO
Molecular Weight: 187.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 88
Melting point (°C): 66
Other Product Names / Synonyms: 4-Bromo-2-methylphenol
Formula:
C₇H₇BrO MW: 187.04 g/mol Boiling Pt: 88 °C Melting Pt: 66 °C |
MDL Number:
MFCD00055435 CAS Number: 2362-12-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...