3-Bromo-1H-pyrrolo[2,3-b]pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3675-1G B3675-5G
TCB3675-1GEA 107.44 CAD
TCB3675-1G TCB3675-5G
3-Bromo-1H-pyrrolo[2,3-b]pyridine ≥98.0% (by GC, titration analysis)
3-Bromo-1H-pyrrolo[2,3-b]pyridine

CAS Number: 74420-15-8
MDL Number: MFCD02179276
Molecular Formula: C7H5BrN2
Molecular Weight: 197.04
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 188


Other Product Names / Synonyms: 3-Bromo-7-azaindole

Formula: C₇H₅BrN₂
Melting Pt: 188 °C
MDL Number: MFCD02179276
CAS Number: 74420-15-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8975a832e06cdd7b5b05d4cd303ead49f138a05b&vid=8975a832e06cdd7b5b05d4cd303ead49f138a05b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR