5-Bromo-2-ethoxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3673-1G B3673-5G
TCB3673-1GEA 48.98 CAD
TCB3673-1G TCB3673-5G
5-Bromo-2-ethoxypyridine ≥98.0% (by GC, titration analysis)
5-Bromo-2-ethoxypyridine

CAS Number: 55849-30-4
MDL Number: MFCD00234311
Molecular Formula: C7H8BrNO
Molecular Weight: 202.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 107
Melting point (°C): 37
Flash Point (°C): 103

Formula: C₇H₈BrNO
Boiling Pt: 107 °C
Melting Pt: 37 °C
Flash Pt: 103 °C
MDL Number: MFCD00234311
CAS Number: 55849-30-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4408a1c67ca301062f392dd9715d362ea980a514&vid=4408a1c67ca301062f392dd9715d362ea980a514&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR