2-Bromo-6-fluoropyridine ≥98.0% (by GC)

Supplier: TCI America

B3667-1G B3667-5G
TCB3667-1GEA 60.04 CAD
TCB3667-1G TCB3667-5G
2-Bromo-6-fluoropyridine ≥98.0% (by GC)
2-Bromo-6-fluoropyridine

CAS Number: 144100-07-2
MDL Number: MFCD03095258
Molecular Formula: C5H3BrFN
Molecular Weight: 175.99
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 164
Melting point (°C): 32

Formula: C₅H₃BrFN
MW: 175.99
Boiling Pt: 164 °C
Melting Pt: 32 °C
MDL Number: MFCD03095258
CAS Number: 144100-07-2
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9973262af67b5e56bbd218ac8354b3c54d59f56&vid=e9973262af67b5e56bbd218ac8354b3c54d59f56&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR