2-Bromo-5-phenylthiophene ≥97.0%

Supplier: TCI America

B3663-1G B3663-5G
TCB3663-1GEA 169.06 CAD
TCB3663-1G TCB3663-5G
2-Bromo-5-phenylthiophene ≥97.0%
2-Bromo-5-phenylthiophene

CAS Number: 29488-24-2
MDL Number: MFCD00159575
Molecular Formula: C10H7BrS
Molecular Weight: 239.13
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 85

Formula: C₁₀H₇BrS
MW: 239.14
Melting Pt: 85 °C
MDL Number: MFCD00159575
CAS Number: 29488-24-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10521889bd8767eb9b0d3e35b60b76a98f09fe61&vid=10521889bd8767eb9b0d3e35b60b76a98f09fe61&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR