(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine ≥96.0% (by GC)

Supplier: TCI America

Synonyms: tert-Butyl (2S,4R)-4-hydroxy-2-hydroxymethylpyrrolidine-1-carboxylate, N-Boc-trans-4-hydroxy-L-prolinol, (2S,4R)-1-Boc-4-hydroxy-2-(hydroxymethyl)pyrrolidine, Boc-Hyp-OL

B3662-1G B3662-5G
TCB3662-1GEA 69.52 CAD
TCB3662-1G TCB3662-5G
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine ≥96.0% (by GC)
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine

CAS Number: 61478-26-0
MDL Number: MFCD02094386
Molecular Formula: C10H19NO4
Molecular Weight: 217.27
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 84
Specific rotation [a]20/D: -45 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (2S,4R)-1-Boc-4-hydroxy-2-(hydroxymethyl)pyrrolidine

Formula: C₁₀H₁₉NO₄
MW: 217.27 g/mol
Melting Pt: 84 °C
MDL Number: MFCD02094386
CAS Number: 61478-26-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9415e9966ca510af76f865a165f904e6e8f604c9&vid=9415e9966ca510af76f865a165f904e6e8f604c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR