(S)-3-(Boc-amino)piperidine ≥98.0%

Supplier: TCI America

Synonyms: (S)-3-Boc-aminopiperidine, (S)-tert-Butyl piperidin-3-ylcarbamate, tert-butyl N-[(3S)-piperidin-3-yl]carbamate

B3660-1G B3660-5G
TCB3660-1GEA 55.3 CAD
TCB3660-1G TCB3660-5G
(S)-3-(Boc-amino)piperidine ≥98.0%
(S)-3-(Boc-amino)piperidine

CAS Number: 216854-23-8
MDL Number: MFCD03093383
Molecular Formula: C10H20N2O2
Molecular Weight: 200.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 121
Specific rotation [a]20/D: -14 deg (C=1, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-3-(Boc-amino)piperidine

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Melting Pt: 121 °C
MDL Number: MFCD03093383
CAS Number: 216854-23-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f107fc4db157eb60e29bc2c43a55e17e21fd020&vid=7f107fc4db157eb60e29bc2c43a55e17e21fd020&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR