4-Bromo-m-cresol ≥98.0%
Supplier: TCI America
Synonyms:
1-Bromo-4-hydroxy-2-methylbenzene, 4-Bromo-3-methylphenol
B3658-25G
B3658-5G
TCB3658-25GEA
290.72
CAD
TCB3658-25G
TCB3658-5G
4-Bromo-m-cresol ≥98.0%
4-Bromo-m-cresol
CAS Number: 14472-14-1
MDL Number: MFCD00079723
Molecular Formula: C7H7BrO
Molecular Weight: 187.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 145
Melting point (°C): 68
Other Product Names / Synonyms: 4-Bromo-m-cresol
Formula:
C₇H₇BrO MW: 187.04 Boiling Pt: 145 °C Melting Pt: 63 |
MDL Number:
MFCD00079723 CAS Number: 14472-14-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...