2-Bromo-5-hydroxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Bromo-5-pyridinol, 6-Bromopyridin-3-ol

B3657-1G B3657-5G
TCB3657-1GEA 30.56 CAD
TCB3657-1G TCB3657-5G
2-Bromo-5-hydroxypyridine ≥98.0% (by GC, titration analysis)
2-Bromo-5-hydroxypyridine

CAS Number: 55717-45-8
MDL Number: MFCD04114184
Molecular Formula: C5H4BrNO
Molecular Weight: 174.00
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 138


Other Product Names / Synonyms: 6-Bromo-3-pyridinol

Formula: C₅H₄BrNO
MW: 174 g/mol
Melting Pt: 138 °C
MDL Number: MFCD04114184
CAS Number: 55717-45-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc62f20f923c374ef7f8d9ab7e9ee9d2dc84e1ca&vid=bc62f20f923c374ef7f8d9ab7e9ee9d2dc84e1ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR