2-(4-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

Synonyms: 4-Bromo-1-(2,3-dihydro-1H-naphtho[1,8-de]-1,3,2-diazaborinyl)benzene, 2-(4-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine

B3655-1G B3655-5G
TCB3655-1GEA 55.3 CAD
TCB3655-1G TCB3655-5G
2-(4-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine ≥98.0% (by GC, total nitrogen)
2-(4-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine

CAS Number: 927384-44-9
MDL Number: MFCD16038141
Molecular Formula: C16H12BBrN2
Molecular Weight: 323.00
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 4-Bromo-1-(2,3-dihydro-1H-naphtho[1,8-de]-1,3,2-diazaborinyl)benzene, 2-(4-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine

Formula: C₁₆H₁₂BBrN₂
MW: 323.00 g/mol
MDL Number: MFCD16038141
CAS Number: 927384-44-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a03a45cbd9081300ae2e5d144a029e2c93c0be58&vid=a03a45cbd9081300ae2e5d144a029e2c93c0be58&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR