2-(3-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine ≥98.0%

Supplier: TCI America

Synonyms: 3-Bromo-1-(2,3-dihydro-1H-naphtho[1,8-de]-1,3,2-diazaborinyl)benzene, 2-(3-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine

B3654-1G B3654-5G
TCB3654-1GEA 105.86 CAD
TCB3654-1G TCB3654-5G
2-(3-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine ≥98.0%
2-(3-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine

CAS Number: 927384-43-8
MDL Number: MFCD16038145
Molecular Formula: C16H12BBrN2
Molecular Weight: 323.00
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 3-Bromo-1-(2,3-dihydro-1H-naphtho[1,8-de]-1,3,2-diazaborinyl)benzene, 2-(3-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine

Formula: C₁₆H₁₂BBrN₂
MDL Number: MFCD16038145
CAS Number: 927384-43-8

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR