2-(2-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

Synonyms: 2-(2-Bromophenyl)-2,3-dihydro-1h-naphtho[1,8-de][1,3,2]diazaborine, 2-Bromo-1-(2,3-dihydro-1H-naphtho[1,8-de]-1,3,2-diazaborinyl)benzene, 2-(2-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine

B3653-1G B3653-5G
TCB3653-1GEA 155.31 CAD
TCB3653-1G TCB3653-5G
2-(2-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine ≥98.0% (by GC, total nitrogen)
2-(2-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine

CAS Number: 927384-42-7
MDL Number: MFCD16038144
Molecular Formula: C16H12BBrN2
Molecular Weight: 323.00
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 2-Bromo-1-(2,3-dihydro-1H-naphtho[1,8-de]-1,3,2-diazaborinyl)benzene, 2-(2-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine

Formula: C₁₆H₁₂BBrN₂
MW: 323.00 g/mol
MDL Number: MFCD16038144
CAS Number: 927384-42-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb91f0d26c7c3f7b7bf09b9a0c88de6d618765d5&vid=cb91f0d26c7c3f7b7bf09b9a0c88de6d618765d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR