9,9'-Bifluorenyl ≥98.0% (by HPLC)
Supplier: TCI America
B3652-1G
B3652-5G
TCB3652-1GEA
143.78
CAD
TCB3652-1G
TCB3652-5G
9,9'-Bifluorenyl ≥98.0% (by HPLC)
9,9'-Bifluorenyl
CAS Number: 1530-12-7
MDL Number: MFCD00001132
Molecular Formula: C26H18
Molecular Weight: 330.43
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 246
Other Product Names / Synonyms: 9,9'-Bifluorene
Formula:
C₂₆H₁₈ Melting Pt: 246 °C |
MDL Number:
MFCD00001132 CAS Number: 1530-12-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...