Boc-N-methyl-D-alanine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B3651-25G
B3651-5G
TCB3651-25GEA
150.1
CAD
TCB3651-25G
TCB3651-5G
Boc-N-methyl-D-alanine ≥98.0% (by GC, titration analysis)
Boc-N-methyl-D-alanine
CAS Number: 16948-16-6
MDL Number: MFCD00037242
Molecular Formula: C9H17NO4
Molecular Weight: 203.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 93
Specific rotation [a]20/D: -30 deg (C=1, EtOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-N-methyl-L-alanine, Boc-N-Me-Ala-OH
Formula:
C₉H₁₇NO₄ Melting Pt: 93 °C |
MDL Number:
MFCD00037242 CAS Number: 16948-16-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...