N-(tert-Butoxycarbonyl)-N-methyl-D-alanine ≥98.0%

Supplier: TCI America

Synonyms: BOC-N-methyl-D-alanine, (R)-2-((tert-Butoxycarbonyl)(methyl)amino)propanoic acid, Boc-N-Me-D-Ala-OH, N-Boc-N-methyl-D-alanine

B3650-1G B3650-5G
TCB3650-1GEA 50.56 CAD
TCB3650-1G TCB3650-5G
N-(tert-Butoxycarbonyl)-N-methyl-D-alanine ≥98.0%
N-(tert-Butoxycarbonyl)-N-methyl-D-alanine

CAS Number: 19914-38-6
MDL Number: MFCD00063137
Molecular Formula: C9H17NO4
Molecular Weight: 203.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 92
Specific rotation [a]20/D: 30 deg (C=1, EtOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-N-methyl-D-alanine, Boc-N-Me-D-Ala-OH

Formula: C₉H₁₇NO₄
MW: 203.24 g/mol
Melting Pt: 92 °C
MDL Number: MFCD00063137
CAS Number: 19914-38-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d94bf993f30e5ce29d176ce6018322dd32aa45aa&vid=d94bf993f30e5ce29d176ce6018322dd32aa45aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR