Buflomedil hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 4-(Pyrrolidin-1-yl)-1-(2,4,6-trimethoxyphenyl)butan-1-one hydrochloride, 4-(1-Pyrrolidinyl)-1-(2,4,6-trimethoxyphenyl)-1-butanone Hydrochloride, Buflomedil HCl

B3647-5G B3647-1G
TCB3647-5GEA 178.54 CAD
TCB3647-5G TCB3647-1G
Buflomedil hydrochloride ≥98.0%
Buflomedil hydrochloride

CAS Number: 35543-24-9
MDL Number: MFCD00078965
Molecular Formula: C17H25NO4
Molecular Weight: 343.85
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 193


Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 4-(1-Pyrrolidinyl)-1-(2,4,6-trimethoxyphenyl)-1-butanone Hydrochloride

Formula: C₁₇H₂₅NO₄
MW: 343.85 g/mol
Melting Pt: 193 °C
MDL Number: MFCD00078965
CAS Number: 35543-24-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfa4ad3c59b010b5c8c25a629efde38849f88188&vid=dfa4ad3c59b010b5c8c25a629efde38849f88188&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR