2-Bromo-5-chloroanisole ≥98.0% (by GC)

Supplier: TCI America

B3646-25G B3646-5G
TCB3646-25GEA 204.93 CAD
TCB3646-25G TCB3646-5G
2-Bromo-5-chloroanisole ≥98.0% (by GC)
2-Bromo-5-chloroanisole

CAS Number: 174913-09-8
MDL Number: MFCD03790889
Molecular Formula: C7H6BrClO
Molecular Weight: 221.48
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.63


Other Product Names / Synonyms: 1-Bromo-4-chloro-2-methoxybenzene

Formula: C₇H₆BrClO
Density: 1.63 (20 °C)
MDL Number: MFCD03790889
CAS Number: 174913-09-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17a45a64eff877bb86b2449a4b83deea4bc380a6&vid=17a45a64eff877bb86b2449a4b83deea4bc380a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR