9,9-Bis(4-bromophenyl)fluorene ≥98.0%

Supplier: TCI America

Synonyms: 9,9-Bis(4-bromophenyl)-9H-fluorene

B3645-1G B3645-5G
TCB3645-1GEA 304.15 CAD
TCB3645-1G TCB3645-5G
9,9-Bis(4-bromophenyl)fluorene ≥98.0%
9,9-Bis(4-bromophenyl)fluorene

CAS Number: 128406-10-0
Molecular Formula: C25H16Br2
Molecular Weight: 476.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 158

Formula: C₂₅H₁₆Br₂
MW: 476.21 g/mol
Melting Pt: 158 °C
MDL Number: MFCD19707068
CAS Number: 128406-10-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=516097e4781ddfe08a8237eb59a0e2299c0f750a&vid=516097e4781ddfe08a8237eb59a0e2299c0f750a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR