1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: 1-(tert-butoxycarbonyl)piperidine-2-carboxylic acid, 1-n-boc-2-piperidinecarboxylic acid, N-BOC-2-Piperidinecarboxylic acid

B3644-25G B3644-5G
TCB3644-25GEA 224.36 CAD
TCB3644-25G TCB3644-5G
1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid ≥98.0%
1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid

CAS Number: 98303-20-9
MDL Number: MFCD01862877
Molecular Formula: C11H19NO4
Molecular Weight: 229.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 131


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 1-Boc-pipecolic Acid, 1-(tert-Butoxycarbonyl)pipecolinic Acid, 1-Boc-pipecolinic Acid, 1-(tert-Butoxycarbonyl)pipecolic Acid, 1-Boc-2-piperidinecarboxylic Acid, Boc-DL-Pip-OH

Formula: C₁₁H₁₉NO₄
MW: 229.276 g/mol
Melting Pt: 131 °C
MDL Number: MFCD01862877
CAS Number: 98303-20-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c04264618c515c2b21fa7f03095edc271dc2e4e&vid=7c04264618c515c2b21fa7f03095edc271dc2e4e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR