1-Bromo-4-propoxybenzene ≥98.0% (by GC)
Supplier: TCI America
B3639-25G
B3639-5G
TCB3639-25GEA
699.94
CAD
TCB3639-25G
TCB3639-5G
1-Bromo-4-propoxybenzene ≥98.0% (by GC)
1-Bromo-4-propoxybenzene
CAS Number: 39969-56-7
MDL Number: MFCD00156160
Molecular Formula: C9H11BrO
Molecular Weight: 215.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 240
Flash Point (°C): 110
Specific Gravity (20/20): 1.35
Other Product Names / Synonyms: 1-(4-Bromophenoxy)propane, 4-Bromophenyl Propyl Ether
Formula:
C₉H₁₁BrO Boiling Pt: 240 °C Density: 1.35 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00156160 CAS Number: 39969-56-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...