N-tert-Butylmaleimide ≥98.0% (by GC)

Supplier: TCI America

B3638-1G B3638-5G
TCB3638-1GEA 51.44 CAD
TCB3638-1G TCB3638-5G
N-tert-Butylmaleimide ≥98.0% (by GC)
N-tert-Butylmaleimide

CAS Number: 4144-22-3
MDL Number: MFCD00191885
Molecular Formula: C8H11NO2
Molecular Weight: 153.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 190
Flash Point (°C): 80
Specific Gravity (20/20): 1.07

Formula: C₈H₁₁NO₂
Boiling Pt: 190 °C
Density: 1.07 (20 °C)
Flash Pt: 80 °C
MDL Number: MFCD00191885
CAS Number: 4144-22-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62d37b2869bab992cfc4dd24c1b3fdbd963f0abd&vid=62d37b2869bab992cfc4dd24c1b3fdbd963f0abd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR