4-Bromosalicylaldehyde ≥98.0%

Supplier: TCI America

Synonyms: 4-Bromo-2-hydroxybenzaldehyde

B3637-1G B3637-5G
TCB3637-1GEA 56.88 CAD
TCB3637-1G TCB3637-5G
4-Bromosalicylaldehyde ≥98.0%
4-Bromosalicylaldehyde

CAS Number: 22532-62-3
MDL Number: MFCD06252606
Molecular Formula: C7H5BrO2
Molecular Weight: 201.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 53
Flash Point (°C): 110


Other Product Names / Synonyms: 4-Bromo-2-hydroxybenzaldehyde

Formula: C₇H₅BrO₂
Melting Pt: 53 °C
Flash Pt: 110 °C
MDL Number: MFCD06252606
CAS Number: 22532-62-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8dad4e4ad1c101e877ff084182fcd9ae8e5a7f0e&vid=8dad4e4ad1c101e877ff084182fcd9ae8e5a7f0e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR