2-Bromo-m-toluic acid ≥98.0%

Supplier: TCI America

B3636-25G B3636-5G
TCB3636-25GEA 110.6 CAD
TCB3636-25G TCB3636-5G
2-Bromo-m-toluic acid ≥98.0%
2-Bromo-m-toluic acid

CAS Number: 53663-39-1
MDL Number: MFCD00079721
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 137


Other Product Names / Synonyms: 2-Bromo-m-toluic Acid

Formula: C₈H₇BrO₂
Melting Pt: 137 °C
MDL Number: MFCD00079721
CAS Number: 53663-39-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c278fbdaf7b29351bb4f92b4c169c6345c679a34&vid=c278fbdaf7b29351bb4f92b4c169c6345c679a34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR