N4-Benzoyl-3',5'-O-(1,1,3,3-tetraisopropyl-1,3-disiloxanediyl)cytidine ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America

B3631-1G B3631-5G
TCB3631-1GEA 353.92 CAD
TCB3631-1G TCB3631-5G
N4-Benzoyl-3',5'-O-(1,1,3,3-tetraisopropyl-1,3-disiloxanediyl)cytidine ≥97.0% (by HPLC, total nitrogen)
N4-Benzoyl-3',5'-O-(1,1,3,3-tetraisopropyl-1,3-disiloxanediyl)cytidine

CAS Number: 69304-43-4
Molecular Formula: C28H43N3O7Si2
Molecular Weight: 589.84
Purity/Analysis Method: >97.0% (HPLC,N)
Form: Crystal
Melting point (°C): 101

Formula: C₂₈H₄₃N₃O₇Si₂
MW: 589.84 g/mol
Melting Pt: 101 °C
CAS Number: 69304-43-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=260ca59edf4406fe69d82a9cb5d1003161423dc3&vid=260ca59edf4406fe69d82a9cb5d1003161423dc3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR