1-(4-Bromophenyl)piperazine hydrochloride ≥98.0%

Supplier: TCI America

B3630-1G B3630-5G
TCB3630-1GEA 137.46 CAD
TCB3630-1G TCB3630-5G
1-(4-Bromophenyl)piperazine hydrochloride ≥98.0%
1-(4-Bromophenyl)piperazine hydrochloride

CAS Number: 68104-62-1
MDL Number: MFCD00674134
Molecular Formula: C10H13BrN2
Molecular Weight: 277.59
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal

Formula: C₁₀H₁₄BrClN₂
CAS Number: 68104-62-1

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR