1-tert-Butyl-4-iodobenzene ≥95.0% (by GC)

Supplier: TCI America

B3624-25G B3624-5G
TCB3624-25GEA 583.49 CAD
TCB3624-25G TCB3624-5G
1-tert-Butyl-4-iodobenzene ≥95.0% (by GC)
1-tert-Butyl-4-iodobenzene

CAS Number: 35779-04-5
MDL Number: MFCD00052339
Molecular Formula: C10H13I
Molecular Weight: 260.12
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Flash Point (°C): 113
Specific Gravity (20/20): 1.45

Formula: C₁₀H₁₃I
Boiling Pt: 118 °C
Density: 1.45 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00052339
CAS Number: 35779-04-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce815e98c86261adae892e8b7ec82181027f7827&vid=ce815e98c86261adae892e8b7ec82181027f7827&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR