Bis(phenylacetyl)disulfide ≥98.0%

Supplier: TCI America

Synonyms: Phenylacetyl Disulfide, Phenylacetyl Disulphide, 2-Phenylacetic dithioperoxyanhydride

B3623-25G B3623-5G
TCB3623-25GEA 349.18 CAD
TCB3623-25G TCB3623-5G
Bis(phenylacetyl)disulfide ≥98.0%
Bis(phenylacetyl)disulfide

CAS Number: 15088-78-5
MDL Number: MFCD00513572
Molecular Formula: C16H14O2S2
Molecular Weight: 302.41
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 60


Product Categories: Synthetic Organic Chemistry, Nucleic Acid Synthesis, Analytical Science, Estradiol, etc. (Environmental Endocrine Disruptors)
Other Product Names / Synonyms: Phenylacetyl Disulfide

Formula: C₁₆H₁₄O₂S₂
MW: 302.41 g/mol
Melting Pt: 60 °C
MDL Number: MFCD00513572
CAS Number: 15088-78-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2d5fbbb772d6dd25b02f94cfc890d08cd9e1a58&vid=f2d5fbbb772d6dd25b02f94cfc890d08cd9e1a58&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR