tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B3619-1G B3619-5G
TCB3619-1GEA 58.46 CAD
TCB3619-1G TCB3619-5G
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate ≥98.0% (by HPLC, titration analysis)
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate

CAS Number: 152120-54-2
MDL Number: MFCD01075122
Molecular Formula: C14H22N4O4
Molecular Weight: 310.35
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 90


Product Categories: Synthetic Organic Chemistry, Guanidinylation

Formula: C₁₄H₂₂N₄O₄
Melting Pt: 90 °C
MDL Number: MFCD01863492
CAS Number: 152120-54-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbccc090bb61ff24691ca4e27c8511cd2903d03e&vid=bbccc090bb61ff24691ca4e27c8511cd2903d03e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR