tert-Butyl-N-allylcarbamate ≥98.0%

Supplier: TCI America

Synonyms: N-Boc-allylamine, N-(tert-Butoxycarbonyl)allylamine, N-Allylcarbamic Acid tert-Butyl Ester, tert-Butyl allylcarbamate

B3618-25G B3618-5G
TCB3618-25GEA 229.1 CAD
TCB3618-25G TCB3618-5G
tert-Butyl-N-allylcarbamate ≥98.0%
tert-Butyl-N-allylcarbamate

CAS Number: 78888-18-3
MDL Number: MFCD00191870
Molecular Formula: C8H15NO2
Molecular Weight: 157.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 49
Melting point (°C): 39
Flash Point (°C): 79


Other Product Names / Synonyms: N-Boc-allylamine, N-(tert-Butoxycarbonyl)allylamine, N-Allylcarbamic Acid tert-Butyl Ester

Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Boiling Pt: 49 °C
Melting Pt: 39 °C
Flash Pt: 79 °C
MDL Number: MFCD00191870
CAS Number: 78888-18-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55d007c406d4aae11aa047c2e01c104ba5d94577&vid=55d007c406d4aae11aa047c2e01c104ba5d94577&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR