N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-D-alanine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
(R)-alpha-(Boc-amino)-2-naphthalenepropionic Acid, Boc-D-2-Nal-OH, (R)-alpha-tert-Butoxycarbonylamino-2-naphthalenepropionic Acid, N-Boc-3-(2-naphthyl)-D-alanine
CAS Number: 76985-10-9
MDL Number: MFCD00076900
Molecular Formula: C18H21NO4
Molecular Weight: 315.37
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 95
Specific rotation [a]20/D: -45 deg (C=1, EtOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-3-(2-naphthyl)-D-alanine, (R)-alpha-tert-Butoxycarbonylamino-2-naphthalenepropionic Acid, (R)-alpha-(Boc-amino)-2-naphthalenepropionic Acid
Formula:
C₁₈H₂₁NO₄ MW: 315.37 g/mol Melting Pt: 95 °C |
MDL Number:
MFCD00076900 CAS Number: 76985-10-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...