N-(tert-Butoxycarbonyl)-4-chloro-L-phenylalanine ≥98.0%

Supplier: TCI America

Synonyms: Boc-Phe(4-Cl)-OH, N-Boc-4-chloro-L-phenylalanine

B3614-1G B3614-5G
TCB3614-1GEA 55.3 CAD
TCB3614-1G TCB3614-5G
N-(tert-Butoxycarbonyl)-4-chloro-L-phenylalanine ≥98.0%
N-(tert-Butoxycarbonyl)-4-chloro-L-phenylalanine

CAS Number: 68090-88-0
MDL Number: MFCD00057808
Molecular Formula: C14H18ClNO4
Molecular Weight: 299.75
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 112
Specific rotation [a]20/D: 24 deg (C=1, EtOAc)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-4-chloro-L-phenylalanine, Boc-Phe(4-Cl)-OH

Formula: C₁₄H₁₈ClNO₄
MW: 299.75 g/mol
Melting Pt: 112 °C
MDL Number: MFCD00057808
CAS Number: 68090-88-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59ffdac971f8ea19186ed108b836a2f869c32d72&vid=59ffdac971f8ea19186ed108b836a2f869c32d72&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR