2-Bromo-6-chloropyridine ≥98.0%

Supplier: TCI America

B3609-1G B3609-5G
TCB3609-1GEA 52.14 CAD
TCB3609-1G TCB3609-5G
2-Bromo-6-chloropyridine ≥98.0%
2-Bromo-6-chloropyridine

CAS Number: 5140-72-7
MDL Number: MFCD00181262
Molecular Formula: C5H3BrClN
Molecular Weight: 192.44
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 90

Formula: C₅H₃BrClN
MW: 192.44
Melting Pt: 90 °C
MDL Number: MFCD00181262
CAS Number: 5140-72-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de4e1462f557082b0dedaeb02767f425bfc71ea9&vid=de4e1462f557082b0dedaeb02767f425bfc71ea9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR