3-Bromo-5-chloropyridine ≥98.0% (by GC)

Supplier: TCI America

B3608-1G B3608-5G
TCB3608-1GEA 74.26 CAD
TCB3608-1G TCB3608-5G
3-Bromo-5-chloropyridine ≥98.0% (by GC)
3-Bromo-5-chloropyridine

CAS Number: 73583-39-8
MDL Number: MFCD04114221
Molecular Formula: C5H3BrClN
Molecular Weight: 192.44
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 80

Formula: C₅H₃BrClN
MW: 192.44 g/mol
Melting Pt: 80 °C
MDL Number: MFCD04114221
CAS Number: 73583-39-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b42868d08b2ce5685d68fc73615397bc0dfd236&vid=6b42868d08b2ce5685d68fc73615397bc0dfd236&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR