2-Bromo-4-nitrobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3607-25G B3607-5G
TCB3607-25GEA 319 CAD
TCB3607-25G TCB3607-5G
2-Bromo-4-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
2-Bromo-4-nitrobenzoic acid

CAS Number: 16426-64-5
Molecular Formula: C7H4BrNO4
Molecular Weight: 246.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 169

Formula: C₇H₄BrNO₄
Melting Pt: 169 °C
MDL Number: MFCD00234252
CAS Number: 16426-64-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eca56ccea2148faef0c4d06ac886a0807abbded9&vid=eca56ccea2148faef0c4d06ac886a0807abbded9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR