N,N'-Bis-Z-1-guanylpyrazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B3605-1G B3605-5G
TCB3605-1GEA 85.32 CAD
TCB3605-1G TCB3605-5G
N,N'-Bis-Z-1-guanylpyrazole ≥98.0% (by HPLC, titration analysis)
N,N'-Bis-Z-1-guanylpyrazole

CAS Number: 152120-55-3
MDL Number: MFCD02683516
Molecular Formula: C20H18N4O4
Molecular Weight: 378.39
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 93


Product Categories: Synthetic Organic Chemistry, Guanidinylation

Formula: C20H18N4O4
Melting Pt: 93 °C
MDL Number: MFCD02683516
CAS Number: 152120-55-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4721f74639758c38a1b2210e83839475845bcbd&vid=e4721f74639758c38a1b2210e83839475845bcbd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR