(±)-1-Boc-3-hydroxypyrrolidine ≥98.0% (by GC)

Supplier: TCI America

B3603-25G B3603-5G
TCB3603-25GEA 474.32 CAD
TCB3603-25G TCB3603-5G
(±)-1-Boc-3-hydroxypyrrolidine ≥98.0% (by GC)
(±)-1-Boc-3-hydroxypyrrolidine

CAS Number: 103057-44-9
MDL Number: MFCD04038535
Molecular Formula: C9H17NO3
Molecular Weight: 187.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 61
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1-Boc-3-pyrrolidinol, 1-(tert-Butoxycarbonyl)-3-hydroxypyrrolidine, 1-Boc-3-hydroxypyrrolidine

Formula: C₉H₁₇NO₃
Melting Pt: 61 °C
MDL Number: MFCD04038535
CAS Number: 103057-44-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db17a55845d3087df8a3a8f4c94e9544190010ee&vid=db17a55845d3087df8a3a8f4c94e9544190010ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR