4-(Benzyloxy)-3-methoxybenzaldehyde ≥98.0% (by GC)

Supplier: TCI America

B3599-25G B3599-5G
TCB3599-25GEA 150.1 CAD
TCB3599-25G TCB3599-5G
4-(Benzyloxy)-3-methoxybenzaldehyde ≥98.0% (by GC)
4-(Benzyloxy)-3-methoxybenzaldehyde

CAS Number: 2426-87-1
MDL Number: MFCD00003365
Molecular Formula: C15H14O3
Molecular Weight: 242.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 185
Melting point (°C): 63

Formula: C₁₅H₁₄O₃
Boiling Pt: 185 °C
Melting Pt: 63 °C
MDL Number: MFCD00003365
CAS Number: 2426-87-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f0d8d77d06bc7b64c5fcd913e3443871b767f09&vid=2f0d8d77d06bc7b64c5fcd913e3443871b767f09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR