4-Bromo-2,2-diphenylbutyric acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
B3597-25G
B3597-5G
TCB3597-25GEA
312.84
CAD
TCB3597-25G
TCB3597-5G
4-Bromo-2,2-diphenylbutyric acid ≥98.0% (by titrimetric analysis)
4-Bromo-2,2-diphenylbutyric acid
CAS Number: 37742-98-6
MDL Number: MFCD01321300
Molecular Formula: C16H15BrO2
Molecular Weight: 319.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 127
Formula:
C₁₆H₁₅BrO₂ MW: 319.20 g/mol Melting Pt: 127 °C |
MDL Number:
MFCD01321300 CAS Number: 37742-98-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...