(R)-3-(Boc-amino)piperidine ≥98.0%

Supplier: TCI America

Synonyms: tert-Butyl N-[(3R)-piperidin-3-yl]carbamate, (R)-3-Boc-aminopiperidine, (R)-tert-Butyl piperidin-3-ylcarbamate

B3591-1G B3591-5G
TCB3591-1GEA 74.26 CAD
TCB3591-1G TCB3591-5G
(R)-3-(Boc-amino)piperidine ≥98.0%
(R)-3-(Boc-amino)piperidine

CAS Number: 309956-78-3
MDL Number: MFCD03093382
Molecular Formula: C10H20N2O2
Molecular Weight: 200.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 123
Specific rotation [a]20/D: 15 deg (C=1, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-3-(Boc-amino)piperidine

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Melting Pt: 123 °C
MDL Number: MFCD03093382
CAS Number: 309956-78-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74def6a8008c064036015bbfff3c6305ce1fdd00&vid=74def6a8008c064036015bbfff3c6305ce1fdd00&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR