tert-Butyl phenyl carbonate ≥96.0% (by HPLC)

Supplier: TCI America

B3590-25G B3590-5G
TCB3590-25GEA 146.94 CAD
TCB3590-25G TCB3590-5G
tert-Butyl phenyl carbonate ≥96.0% (by HPLC)
tert-Butyl phenyl carbonate

CAS Number: 6627-89-0
MDL Number: MFCD00008804
Molecular Formula: C11H14O3
Molecular Weight: 194.23
Purity/Analysis Method: >96.0% (HPLC)
Form: Clear Liquid
Boiling point (°C): 78
Flash Point (°C): 101
Specific Gravity (20/20): 1.05


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: Carbonic Acid tert-Butyl Phenyl Ester

Formula: C₁₁H₁₄O₃
Boiling Pt: 78 °C
Density: 1.05 (20 °C)
Flash Pt: 101 °C
MDL Number: MFCD00008804
CAS Number: 6627-89-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=901bbc411f47fead78890bdbd947f3fd6f3a22b5&vid=901bbc411f47fead78890bdbd947f3fd6f3a22b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR