tert-Butyl-3-oxopyrrolidine-1-carboxylate ≥95.0% (by GC)

Supplier: TCI America

B3589-1G B3589-5G
TCB3589-1GEA 26.86 CAD
TCB3589-1G TCB3589-5G
tert-Butyl-3-oxopyrrolidine-1-carboxylate ≥95.0% (by GC)
tert-Butyl-3-oxopyrrolidine-1-carboxylate

CAS Number: 101385-93-7
MDL Number: MFCD01631194
Molecular Formula: C9H15NO3
Molecular Weight: 185.22
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 38
Flash Point (°C): 110
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1-Boc-3-pyrrolidone, 1-tert-Butoxycarbonyl-3-pyrrolidinone, 1-Boc-3-pyrrolidinone, tert-Butyl 3-Oxopyrrolidine-1-carboxylate, 3-Oxopyrrolidine-1-carboxylic Acid tert-Butyl Ester

Formula: C₉H₁₅NO₃
Melting Pt: 38 °C
Flash Pt: 110 °C
MDL Number: MFCD01631194
CAS Number: 101385-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=385d11d7993cd5f44785759ac12f2505b1892041&vid=385d11d7993cd5f44785759ac12f2505b1892041&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR