Dithiodiglycol (bis(2-hydroxyethyl) disulfide) ∼50% in water

Supplier: TCI America

B3587-25G B3587-500G
TCB3587-25GEA 52.93 CAD
TCB3587-25G TCB3587-500G
Dithiodiglycol (bis(2-hydroxyethyl) disulfide) ∼50% in water
Dithiodiglycol (Bis(2-hydroxyethyl) disulphide)

CAS Number: 1892-29-1
MDL Number: MFCD00002906
Molecular Formula: C4H10O2S2
Molecular Weight: 154.24
Form: Clear Liquid
Color: Colorless


Other Product Names / Synonyms: 2,2'-Dithiodiethanol, 2-Hydroxyethyl Disulfide

Formula: C₄H₁₀O₂S₂
MDL Number: MFCD00002906
CAS Number: 1892-29-1
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f4ddbde9321f90c2482b76042803a4c0609a21e&vid=5918e86c8f3a6e3b9677ceed6aba0b67592dfd3e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR