1,3,5-Benzenetrimethanol ≥95.0%

Supplier: TCI America

Synonyms: 1,3,5-Tris(hydroxymethyl)benzene, Benzene-1,3,5-triyltrimethanol

B3586-1G B3586-5G
TCB3586-1GEA 213.3 CAD
TCB3586-1G TCB3586-5G
1,3,5-Benzenetrimethanol ≥95.0%
1,3,5-Benzenetrimethanol

CAS Number: 4464-18-0
MDL Number: MFCD01108484
Molecular Formula: C9H12O3
Molecular Weight: 168.19
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 78


Product Categories: Materials Chemistry, Dendrimer Building Blocks
Other Product Names / Synonyms: 1,3,5-Tris(hydroxymethyl)benzene

Formula: C₉H₁₂O₃
Melting Pt: 78 °C
MDL Number: MFCD01108484
CAS Number: 4464-18-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72a0fb0ca3889f79776c6e0c18f16d786d4b90d5&vid=72a0fb0ca3889f79776c6e0c18f16d786d4b90d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR