Tetrazolium blue chloride ≥95.0% (by titrimetric analysis) for biochemical research

Supplier: TCI America

B3581-1G B3581-5G
TCB3581-1GEA 66.04 CAD
TCB3581-1G TCB3581-5G
Tetrazolium blue chloride ≥95.0% (by titrimetric analysis) for biochemical research
Tetrazolium blue chloride

[for Biochemical Research]
CAS Number: 1871-22-3
MDL Number: MFCD00040933
Molecular Formula: C40H32Cl2N8O2
Molecular Weight: 727.65
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 248


Product Categories: Bioscience, Stains & Dyes, Alkali Phosphatase Substrates
Other Product Names / Synonyms: Ditetrazolium Chloride, Tetrazolium Blue

Formula: C₄₀H₃₂Cl₂N₈O₂
Melting Pt: 248 °C
MDL Number: MFCD00040933
CAS Number: 1871-22-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bdfd8efc013dee1b385135f8097733132479b0e&vid=3c5270072e3c952af374a5cc90e6e1b4c191384e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR