1,4-Bis(butyryloxy)-2-butene ≥95.0%

Supplier: TCI America

Synonyms: 2-Butene-1,4-diol Dibutyrate

B3580-25G B3580-5G
TCB3580-25GEA 223.1 CAD
TCB3580-25G TCB3580-5G
1,4-Bis(butyryloxy)-2-butene ≥95.0%
1,4-Bis(butyryloxy)-2-butene

CAS Number: 1572-84-5
MDL Number: MFCD07368634
Molecular Formula: C12H20O4
Molecular Weight: 228.29
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 2-Butene-1,4-diol Dibutyrate

Formula: C₁₂H₂₀O₄
MW: 228.29 g/mol
Boiling Pt: 155 °C
Density: 1.00 (20 °C)
MDL Number: MFCD07368634
CAS Number: 1572-84-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b8f052335996606dd35ba4dfef69d5895affa50&vid=5b8f052335996606dd35ba4dfef69d5895affa50&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR