5-Bromo-2-fluoro-4-methylpyridine ≥98.0% (by GC)
Supplier: TCI America
B3579-1G
B3579-5G
TCB3579-1GEA
28.44
CAD
TCB3579-1G
TCB3579-5G
5-Bromo-2-fluoro-4-methylpyridine ≥98.0% (by GC)
5-Bromo-2-fluoro-4-methylpyridine
CAS Number: 864830-16-0
MDL Number: MFCD09839213
Molecular Formula: C6H5BrFN
Molecular Weight: 190.02
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.62
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Bromo-2-fluoro-4-picoline
Formula:
C₆H₅BrFN Density: 1.62 (20 °C) |
MDL Number:
MFCD09839213 CAS Number: 864830-16-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...