11-Bromo-1-undecene ≥94.0% (by GC)

Supplier: TCI America

B3576-5G B3576-25G
TCB3576-5GEA 104.28 CAD
TCB3576-5G TCB3576-25G
11-Bromo-1-undecene ≥94.0% (by GC)
11-Bromo-1-undecene

CAS Number: 7766-50-9
MDL Number: MFCD00040825
Molecular Formula: C11H21Br
Molecular Weight: 233.19
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 113
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: 10-Undecenyl Bromide

Formula: C₁₁H₂₁Br
Boiling Pt: 115 °C
Density: 1.08 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00040825
CAS Number: 7766-50-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f64b36e21a91e59c68439e5bcfda69f7e761bc9&vid=6f64b36e21a91e59c68439e5bcfda69f7e761bc9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR