Benzyl phenyl carbonate ≥95.0% (by GC)

Supplier: TCI America

B3574-25G B3574-5G
TCB3574-25GEA 243.32 CAD
TCB3574-25G TCB3574-5G
Benzyl phenyl carbonate ≥95.0% (by GC)
Benzyl phenyl carbonate

CAS Number: 28170-07-2
MDL Number: MFCD07784341
Molecular Formula: C14H12O3
Molecular Weight: 228.25
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 131
Specific Gravity (20/20): 1.16


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: Carbonic Acid Benzyl Phenyl Ester

Formula: C₁₄H₁₂O₃
Boiling Pt: 131 °C
Density: 1.16 (20 °C)
MDL Number: MFCD07784341
CAS Number: 28170-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab98ba615d905b94ea2f106a2ea66fd984cf6d07&vid=ab98ba615d905b94ea2f106a2ea66fd984cf6d07&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR