4,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole ≥95.0%

Supplier: TCI America

Synonyms: 2,1,3-Benzothiadiazole-4,7-diboronic Acid Bis(pinacol) Ester, 4,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[c][1,2,5]thiadiazole

B3573-1G B3573-5G
TCB3573-1GEA 385.52 CAD
TCB3573-1G TCB3573-5G
4,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole ≥95.0%
4,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole

CAS Number: 934365-16-9
Molecular Formula: C18H26B2N2O4S
Molecular Weight: 388.10
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 214


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives, Materials Chemistry, Building Blocks for Semiconducting Polymers, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 2,1,3-Benzothiadiazole-4,7-diboronic Acid Bis(pinacol) Ester

Formula: C₁₈H₂₆B₂N₂O₄S
MW: 388.10 g/mol
Melting Pt: 214 °C
MDL Number: MFCD11656086
CAS Number: 934365-16-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=529db408efe9712822972a05c2a5776cf5d95a97&vid=529db408efe9712822972a05c2a5776cf5d95a97&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR